accessibility menu, dialog, popup

UserName

CH3NOSOONONNSOON

CAS RN 127191-97-3

IUPAC Name: 4-[2-(N-methylisoquinoline-5-sulfonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl]phenyl isoquinoline-5-sulfonate
Synonyms: 4-[2-(N-methylisoquinoline-5-sulfonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl]phenyl isoquinoline-5-sulfonate
Molecular Weight (g/mol): 721.85
Molecular Formula: C38H35N5O6S2
InChi Key: RJVLFQBBRSMWHX-UHFFFAOYNA-N
SMILES: CN(C(CC1=CC=C(OS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1)C(=O)N1CCN(CC1)C1=CC=CC=C1)S(=O)(=O)C1=C2C=CN=CC2=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results