accessibility menu, dialog, popup

UserName

CAS RN 126930-72-1

IUPAC Name: 1-bromo-4-dodecylbenzene
Synonyms: 1-bromo-4-dodecylbenzene
Molecular Weight (g/mol): 325.33
Molecular Formula: C18H29Br
InChi Key: RFFMRBISTZHCNY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=C(Br)C=C1
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)