missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-dodecylbenzene, 98%

Catalog No. AAH3296314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3296314 25 g
AAH3296306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3296314 Supplier Thermo Scientific Chemicals Supplier No. H3296314

CAS: 126930-72-1 | C18H29Br | 325.334 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126930-72-1
Molecular Formula C18H29Br
Molecular Weight (g/mol) 325.334
MDL Number MFCD00191386
InChI Key RFFMRBISTZHCNY-UHFFFAOYSA-N
Synonym 4-dodecylbromobenzene, 1-bromo-4-n-dodecylbenzene, benzene, 1-bromo-4-dodecyl, 4-bromododecylbenzene, pubchem22735, acmc-209bby, 1-bromo-4-dodecyl-benzene, ksc494g6d, 4-n-dodecylbromobenzene
PubChem CID 15120506
IUPAC Name 1-bromo-4-dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)Br

Specifications

Density 1.07
Boiling Point 376°C
Flash Point >110°C (230°F)
Refractive Index 1.5034
Quantity 25 g
Formula Weight 325.33
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-n-dodecylbenzene

RUO – Research Use Only