accessibility menu, dialog, popup

UserName

H3COPOOCH3OCH3

CAS RN 126-73-8

IUPAC Name: tributyl phosphate
Synonyms: tributyl phosphate tributyl phosphate (substance) disflamoll TB tributylfosfat tri-n-butylphosphate tri-N-butylphosphate tributylfosfaat tri-(n-butyl )-phosphate tributilfosfato butyl phosphate
Molecular Weight (g/mol): 266.32
Molecular Formula: C12H27O4P
InChi Key: STCOOQWBFONSKY-UHFFFAOYSA-N
SMILES: CCCCOP(=O)(OCCCC)OCCCC
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
  • (10)
  • (3)
Physical Form 
  • (16)
Grade 
  • (13)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (10)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (10)
  • (3)
  • (16)
  • (13)
  • (3)
  • (3)
  • (3)