accessibility menu, dialog, popup

UserName

CH3NNNNNHNFFF

CAS RN 1255517-76-0

IUPAC Name: 2-{[4-(5-ethylpyrimidin-4-yl)piperazin-1-yl]methyl}-6-(trifluoromethyl)-1H-1,3-benzodiazole
Synonyms: 2-{[4-(5-ethylpyrimidin-4-yl)piperazin-1-yl]methyl}-5-(trifluoromethyl)-3H-1,3-benzodiazole
Molecular Weight (g/mol): 390.41
Molecular Formula: C19H21F3N6
InChi Key: FBLPQCAQRNSVHB-UHFFFAOYSA-N
SMILES: CCC1=CN=CN=C1N1CCN(CC2=NC3=CC=C(C=C3N2)C(F)(F)F)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results