accessibility menu, dialog, popup

UserName

H2OH2OFeHOOHOHOHOHOOHOOHOHOHOHOO

CAS RN 12389-15-0

IUPAC Name: λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate
Synonyms: λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate
Molecular Weight (g/mol): 482.17
Molecular Formula: C12H26FeO16
InChi Key: OKGNXSFAYMSVNN-UHFFFAOYNA-L
SMILES: O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O
Grade 
  • (4)
  • (4)
Percent Purity 
  • (8)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (4)
  • (4)
  • (8)
  • (2)
  • (2)
  • (2)
  • (2)