accessibility menu, dialog, popup

UserName

H3CH3CCH3PHNCH3CH3CH3H3CH2CCH2ClPd

CAS RN 1235509-04-2

IUPAC Name: chloropalladiumylium; prop-2-en-1-ide; {[4-(di-tert-butylphosphanyl)phenyl]methyl}(methyl)amine
Synonyms: chloropalladiumylium; prop-2-en-1-ide; {[4-(di-tert-butylphosphanyl)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol): 448.32
Molecular Formula: C19H33ClNPPd
InChi Key: OSTWSTKYPFRHAA-UHFFFAOYSA-M
SMILES: Cl[Pd+].[CH2-]C=C.CNCC1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results