missing translation for 'onlineSavingsMsg'
Learn More

Allyl(chloro)[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)

Catalog No. AA4633203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4633203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4633203 Supplier Thermo Scientific Chemicals Supplier No. 04633203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1235509-04-2 | C19H33ClNPPd | 448.324 g/mol

The derivative of this compound serves as an exceptionally mild and versatile catalyst for Heck reactions of aryl chlorides and bromides. A sterically and electronically diverse array of aryl bromides, as well as activated aryl chlorides, couple with a range of mono- and disubstituted olefins at room temperature, furnishing the arylated product with high E/Z stereoselection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1235509-04-2
Molecular Formula C19H33ClNPPd
Molecular Weight (g/mol) 448.324
MDL Number MFCD25372543
InChI Key OSTWSTKYPFRHAA-UHFFFAOYSA-M
Synonym pdclallyl amphos, allyl chloro di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii
PubChem CID 73994976
IUPAC Name chloropalladium(1+);1-(4-ditert-butylphosphanylphenyl)-N-methylmethanamine;prop-1-ene
SMILES CC(C)(C)P(C1=CC=C(C=C1)CNC)C(C)(C)C.[CH2-]C=C.Cl[Pd+]

Specifications

Color Yellow
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 448.32
Physical Form Solid
Chemical Name or Material Allyl (chloro)[di-tert-butyl (4-dimethylaminophenyl)phosphine]palladium (II)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only