accessibility menu, dialog, popup

UserName

Check Out the New Spotlight Offers and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 123334-18-9

AsH2OHClCl

CAS RN 123334-18-9

IUPAC Name: tetraphenylarsanium hydrate hydrochloride chloride
Synonyms: tetraphenyl-arsonium hydrate hydrochloride chloride
Molecular Weight (g/mol): 473.27
Molecular Formula: C24H23AsCl2O
InChi Key: HRCBNZHNCWEFFR-UHFFFAOYSA-M
SMILES: O.Cl.[Cl-].C1=CC=C(C=C1)[As+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.