accessibility menu, dialog, popup

UserName

OOHN

CAS RN 123-56-8

IUPAC Name: pyrrolidine-2,5-dione
Synonyms: succinimide butanimide succinic imide succinic acid imide succinimide-sauba
Molecular Weight (g/mol): 99.09
Molecular Formula: C4H5NO2
InChi Key: KZNICNPSHKQLFF-UHFFFAOYSA-N
SMILES: O=C1CCC(=O)N1
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)