accessibility menu, dialog, popup

UserName

CH3OCH3O

CAS RN 123-54-6

IUPAC Name: pentane-2,4-dione
Synonyms: acetylacetone acetyl-2-propanone ACAC 2-propanone, acetyl- hacac acetyl 2-propanone
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: YRKCREAYFQTBPV-UHFFFAOYSA-N
SMILES: CC(=O)CC(C)=O
Percent Purity 
  • (2)
  • (1)
  • (7)
  • (4)
Boiling Point 
  • (4)
  • (2)
  • (3)
  • (4)
Physical Form 
  • (13)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (1)
  • (4)
  • (2)
  • (7)
  • (2)
  • (1)
  • (7)
  • (4)
  • (4)
  • (2)
  • (3)
  • (4)
  • (13)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)