accessibility menu, dialog, popup

UserName

H3COOSOOCH3

CAS RN 123-28-4

IUPAC Name: dodecyl 3-{[3-(dodecyloxy)-3-oxopropyl]sulfanyl}propanoate
Synonyms: dilauryl thiodipropionate
Molecular Weight (g/mol): 514.85
Molecular Formula: C30H58O4S
InChi Key: GHKOFFNLGXMVNJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results