accessibility menu, dialog, popup

UserName

OHOHOHOHOHOHKKPt

CAS RN 12285-90-4

IUPAC Name: platinum(4+) dipotassium hexahydroxide
Synonyms: platinum(4+) dipotassium hexahydroxide
Molecular Weight (g/mol): 375.32
Molecular Formula: H6K2O6Pt
InChi Key: ALHHYLLHZKAXCW-UHFFFAOYSA-H
SMILES: [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[K+].[K+].[Pt+4]
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results