accessibility menu, dialog, popup

UserName

CAS RN 1220-83-3

IUPAC Name: 4-amino-N-(6-methoxypyrimidin-4-yl)benzene-1-sulfonamide
Synonyms: sulfamonomethoxine sulfamonometoxina sulfamonomethoxin daimeton 磺胺间甲氧嘧啶 sulfamonomethoxinum
Molecular Weight (g/mol): 280.3
Molecular Formula: C11H12N4O3S
InChi Key: WMPXPUYPYQKQCX-UHFFFAOYSA-N
SMILES: COC1=CC(NS(=O)(=O)C2=CC=C(N)C=C2)=NC=N1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)