accessibility menu, dialog, popup

UserName

CAS RN 122-48-5

IUPAC Name: 4-(4-hydroxy-3-methoxyphenyl)butan-2-one
Synonyms: zingerone
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: OJYLAHXKWMRDGS-UHFFFAOYSA-N
SMILES: COC1=CC(CCC(C)=O)=CC=C1O
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)