accessibility menu, dialog, popup

UserName

CAS RN 122-46-3

IUPAC Name: 3-methylphenyl acetate
Synonyms: M-tolylacetate M-methylphenylacetic acid M-cresyl acetate
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: OTGAHJPFNKQGAE-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)