missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyl Acetate 99.0+%, TCI America™

Catalog No. A0659500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0659500G 500 g
A065925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0659500G Supplier TCI America Supplier No. A0659500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-46-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00041910
InChI Key OTGAHJPFNKQGAE-UHFFFAOYSA-N
Synonym m-tolyl acetate, m-cresyl acetate, cresatin, m-acetoxytoluene, m-cresol acetate, acetic acid m-tolyl ester, cresatin-sulzberger, kresatin, acetic acid, 3-methylphenyl ester, m-methylphenyl acetate
PubChem CID 67406
IUPAC Name 3-methylphenyl acetate
SMILES CC(=O)OC1=CC=CC(C)=C1

Specifications

Melting Point 12°C
Color Colorless
Boiling Point 212°C
Quantity 500 g
Formula Weight 150.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Tolyl Acetate
EINECSNumber (3)-0644
TSCA Yes