accessibility menu, dialog, popup

UserName

CAS RN 122-11-2

H3CONHSOONH2NOH3CN

CAS RN 122-11-2

IUPAC Name: 4-amino-N-(2,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide
Synonyms: sulfadimethoxine madrigid madribon sulxin fuxal di-methox abcid (TN) abcid sulfadimethoxine (substance) sulfadimetossina
Molecular Weight (g/mol): 310.33
Molecular Formula: C12H14N4O4S
InChi Key: ZZORFUFYDOWNEF-UHFFFAOYSA-N
SMILES: COC1=CC(NS(=O)(=O)C2=CC=C(N)C=C2)=NC(OC)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results