missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadimethoxine 98.0+%, TCI America™
Supplier: TCI America S035925G
Specifications
| Sulfadimethoxine | |
| 200°C | |
| C12H14N4O4S | |
| 25 g | |
| COC1=NC(=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N)OC | |
| 310.328 | |
| CHEBI:32161 | |
| ≥98.0% (HPLC,T) |
| 122-11-2 | |
| White-Yellow | |
| MFCD00057345 | |
| ZZORFUFYDOWNEF-UHFFFAOYSA-N | |
| 4-amino-N-(2,6-dimethoxypyrimidin-4-yl)benzenesulfonamide | |
| 5323 | |
| 310.33 | |
| Crystalline Powder |
Chemical Identifiers
| 122-11-2 | |
| 310.328 | |
| ZZORFUFYDOWNEF-UHFFFAOYSA-N | |
| CHEBI:32161 | |
| COC1=NC(=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N)OC |
| C12H14N4O4S | |
| MFCD00057345 | |
| 5323 | |
| 4-amino-N-(2,6-dimethoxypyrimidin-4-yl)benzenesulfonamide |
Safety and Handling
RTECSNumber : WO9030000
TSCA : No