accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNN

CAS RN 1219956-23-6

IUPAC Name: 2,4-diphenyl-6-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine
Synonyms: 2,4-diphenyl-6-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,5-triazine
Molecular Weight (g/mol): 435.33
Molecular Formula: C27H26BN3O2
InChi Key: PVGOPEUJUVXCGN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results