accessibility menu, dialog, popup

UserName

CAS RN 1214336-88-5

CH3OONBrF

CAS RN 1214336-88-5

IUPAC Name: methyl 6-bromo-5-fluoropyridine-3-carboxylate
Synonyms: methyl 6-bromo-5-fluoropyridine-3-carboxylate
Molecular Weight (g/mol): 234.02
Molecular Formula: C7H5BrFNO2
InChi Key: PXHKAVXJQLVUBI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(Br)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results