accessibility menu, dialog, popup

UserName

CAS RN 12137-21-2

H2OOPtO

CAS RN 12137-21-2

IUPAC Name: dioxoplatinum hydrate
Synonyms: adams's catalyst hydrate
Molecular Weight (g/mol): 245.1
Molecular Formula: H2O3Pt
InChi Key: SPGAMGILENUIOF-UHFFFAOYSA-N
SMILES: O.O=[Pt]=O
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)