accessibility menu, dialog, popup

UserName

CAS RN 121359-48-6

IUPAC Name: 2-(tributylstannyl)-1,3-thiazole
Synonyms: 2-(tributylstannyl)-1,3-thiazole
Molecular Weight (g/mol): 374.17
Molecular Formula: C15H29NSSn
InChi Key: WUOFQGMXQCSPPV-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)