CAS RN 12125-03-0
CAS RN 12125-03-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Potassium tin(IV) oxide trihydrate, 95%
CAS: 12125-03-0 Molecular Formula: H6K2O6Sn Molecular Weight (g/mol): 298.95 MDL Number: MFCD00150391 InChI Key: HTHDWDSBYOUAFF-UHFFFAOYSA-N Synonym: potassium stannate trihydrate,unii-r38y86o4sy,2k.so3n.3h2o,potassium tin iv oxide trihydrate,dipotassium dioxido oxo tin trihydrate,dipotassium ion oxostannanebis olate trihydrate,dipotassium oxostannanebis olate trihydrate,dipotassium bis oxidanidyl-oxidanylidene-tin trihydrate PubChem CID: 18694711 IUPAC Name: dipotassium;dioxido(oxo)tin;trihydrate SMILES: O.O.O.[K+].[K+].[O-][Sn]([O-])=O