accessibility menu, dialog, popup

UserName

CAS RN 12112-67-3

IUPAC Name: diiridium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
Synonyms: diiridium(1+) bis(1,5-cyclooctadiene, (Z,Z)-) dichloride
Molecular Weight (g/mol): 671.7
Molecular Formula: C16H24Cl2Ir2
InChi Key: ZFOUDQNHNLDNLD-MIXQCLKLSA-L
SMILES: [Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)