accessibility menu, dialog, popup

UserName

PdClCl

CAS RN 12107-56-1

IUPAC Name: palladium(2+) (1Z,5Z)-cycloocta-1,5-diene dichloride
Synonyms: palladium(2+) 1,5-cyclooctadiene, (Z,Z)- dichloride
Molecular Weight (g/mol): 285.5
Molecular Formula: C8H12Cl2Pd
InChi Key: RRHPTXZOMDSKRS-PHFPKPIQSA-L
SMILES: [Cl-].[Cl-].[Pd++].C1C\C=C/CC\C=C/1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)