accessibility menu, dialog, popup

UserName

H3CNCH3

CAS RN 121-69-7

IUPAC Name: N,N-dimethylaniline
Synonyms: dimethylaniline versneller NL 63/10 dwumetyloanilina N,N-(dimethylamino)benzene
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: JLTDJTHDQAWBAV-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (3)
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (4)
  • (7)
Physical Form 
  • (11)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (4)
  • (4)
  • (7)
  • (4)
  • (3)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (7)
  • (11)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)