accessibility menu, dialog, popup

UserName

CAS RN 121-33-5

IUPAC Name: 4-hydroxy-3-methoxybenzaldehyde
Synonyms: vanillin vanillin (product) zimco vanilla methylprotocatechuic aldehyde protocatechualdehyde, methyl- vaniline vanillin (natural) methyl-protocatechualdehyde sodium salt vanillin
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: MWOOGOJBHIARFG-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC=C1O
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (5)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (7)
  • (5)
  • (1)
Grade 
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (5)
  • (1)
  • (5)
  • (3)
  • (8)
  • (1)
  • (2)
  • (2)
  • (5)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (7)
  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)