missing translation for 'onlineSavingsMsg'
Learn More

Vanillin, NF, 97-103%, Spectrum™ Chemical

Catalog No. 18607701
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607701 50 kg Amber Glass Bottle
18607697 125 g Amber Glass Bottle
18607698 500 g Amber Glass Bottle
18607699 2.5 kg Amber Glass Bottle
18607700 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18607701 Supplier Spectrum Chemical Mfg Cor Supplier No. VA13050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VA130, 121-33-5, C8H8O3

Spectrum™ Chemical Vanillin, NF is used as a flavoring agent in pharmaceutical formulations. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-33-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00006942,MFCD08702848
InChI Key MWOOGOJBHIARFG-UHFFFAOYSA-N
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1O

Specifications

CAS Max % 1
Ignition Residue 0.0005
Loss on Drying 0.01
Melting Range 81-83°C
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity 97 to 103%
Grade NF
Physical Form Crystals or Flakes
Chemical Name or Material Vanillin