accessibility menu, dialog, popup

UserName

CAS RN 121-30-2

IUPAC Name: 4-amino-6-chlorobenzene-1,3-disulfonamide
Synonyms: salmid idorese chloaminophenamide C6H8clN3O4S2
Molecular Weight (g/mol): 285.72
Molecular Formula: C6H8ClN3O4S2
InChi Key: IHJCXVZDYSXXFT-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(C=C1S(N)(=O)=O)S(N)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)