missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%

Catalog No. AC251851000
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC251851000 100 g Glass bottle
AC251850050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC251851000 Supplier Thermo Scientific Chemicals Supplier No. 251851000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-30-2 | C6H8ClN3O4S2 | 285.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-30-2
Molecular Formula C6H8ClN3O4S2
Molecular Weight (g/mol) 285.73
InChI Key IHJCXVZDYSXXFT-UHFFFAOYSA-N
Synonym 4-amino-6-chloro-1,3-benzenedisulfonamide, chloraminophenamide, salmid, idorese, chloroaminophenamide, 1,3-benzenedisulfonamide, 4-amino-6-chloro, 5-chloro-2,4-disulfamylaniline, 4-amino-6-chloro-m-benzenedisulfonamide, 3-chloro-4,6-disulfamoylaniline, 5-chloro-2,4-disulfamoylaniline
PubChem CID 67136
ChEBI CHEBI:3602
IUPAC Name 4-amino-6-chlorobenzene-1,3-disulfonamide
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)N)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 260.0°C to 263.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 15,2076
Formula Weight 285.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Amino-6-chloro-1, 3-benzenedisulfonamide
Show More Show Less
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Wear protective gloves/eye protection/face protection.
Rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Get medical attention/advice if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-463-1

RUO – Research Use Only