accessibility menu, dialog, popup

UserName

CH3NOONOO

CAS RN 121-14-2

IUPAC Name: 1-methyl-2,4-dinitrobenzene
Synonyms: 2,4-dinitrotoluene 2,4-dinitromethylbenzene DNT
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: RMBFBMJGBANMMK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results