accessibility menu, dialog, popup

UserName

CAS RN 1207346-29-9

IUPAC Name: oxolane-3-sulfonyl chloride
Synonyms: oxolane-3-sulfonyl chloride
Molecular Weight (g/mol): 170.61
Molecular Formula: C4H7ClO3S
InChi Key: MVZIUOJNVVIOEY-UHFFFAOYNA-N
SMILES: ClS(=O)(=O)C1CCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results