CAS RN 120711-81-1
IUPAC Name:
7-nitro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms:
7-nitro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol):
180.16
Molecular Formula:
C8H8N2O3
InChi Key:
YKCFDUNYLMTXFC-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC2=C(NCCO2)C=C1
1
–
1
of
1
results
1
7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals
CAS: 120711-81-1 Molecular Formula: C8H8N2O3 Molecular Weight (g/mol): 180.163 MDL Number: MFCD11603433 InChI Key: YKCFDUNYLMTXFC-UHFFFAOYSA-N Synonym: 7-nitro-3,4-dihydro-2h-1,4-benzooxazine,7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine,7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine,2,3-dihydro-7-nitro-1,4-benzoxazine,3,4-dihydro-7-nitro-2h-1,4-benzoxazine,7-nitro-2,3-dihydro-4h-1,4-benzoxazine,2h-1,4-benzoxazine, 3,4-dihydro-7-nitro,3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine,7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine PubChem CID: 18416151 IUPAC Name: 7-nitro-3,4-dihydro-2H-1,4-benzoxazine SMILES: C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]