accessibility menu, dialog, popup

UserName

PGa

CAS RN 12063-98-8

IUPAC Name: gallanylidynephosphane
Synonyms: gallium phosphide
Molecular Weight (g/mol): 100.7
Molecular Formula: GaP
InChi Key: HZXMRANICFIONG-UHFFFAOYSA-N
SMILES: P#[Ga]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results