accessibility menu, dialog, popup

UserName

CAS RN 1206161-97-8

OHNNNNNSOO

CAS RN 1206161-97-8

IUPAC Name: N-(5-{4-[(1,1-dioxo-1λ⁶-thiomorpholin-4-yl)methyl]phenyl}-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide
Synonyms: filgotinib
Molecular Weight (g/mol): 425.51
Molecular Formula: C21H23N5O3S
InChi Key: RIJLVEAXPNLDTC-UHFFFAOYSA-N
SMILES: O=C(NC1=NN2C(C=CC=C2C2=CC=C(CN3CCS(=O)(=O)CC3)C=C2)=N1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results