missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Filgotinib, 95%

Catalog No. AC468181000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468181000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468181000 Supplier Thermo Scientific™ Supplier No. 468181000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Filgotinib
CAS 1206161-97-8
Synonym N-[5-[4-[(1,1-Dioxido-4-thiomorpholinyl)methyl]phenyl][1,2,4 ]triazolo[1,5-a]pyridin-2-yl]-cyclopropanecarboxamide
Quantity 100 mg
Molecular Formula C21 H23 N5 O3 S
Formula Weight 425.51
Physical Form Solid
Product Type Reagent
Purity 95%