accessibility menu, dialog, popup

UserName

CAS RN 12044-79-0

IUPAC Name: arsanethiol
Synonyms: arsanethiol
Molecular Weight (g/mol): 110.01
Molecular Formula: AsH3S
InChi Key: BMOFEAKQXGGUPE-UHFFFAOYSA-N
SMILES: S[AsH2]
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results