accessibility menu, dialog, popup

UserName

ORuO

CAS RN 12036-10-1

IUPAC Name: dioxoruthenium
Synonyms: ruthenium(IV) oxide
Molecular Weight (g/mol): 133.07
Molecular Formula: O2Ru
InChi Key: WOCIAKWEIIZHES-UHFFFAOYSA-N
SMILES: O=[Ru]=O
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)