CAS RN 12030-24-9
CAS RN 12030-24-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Indium(III) sulfide, 99.995% (metals basis)
CAS: 12030-24-9 Molecular Formula: In2S3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011061 InChI Key: SIXIBASSFIFHDK-UHFFFAOYSA-N IUPAC Name: diindium(3+) trisulfanediide SMILES: [S--].[S--].[S--].[In+3].[In+3]
2
Indium(III) sulfide, 99.999% (metals basis)
CAS: 12030-24-9 Molecular Formula: In2S3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011061 InChI Key: SIXIBASSFIFHDK-UHFFFAOYSA-N IUPAC Name: diindium(3+) trisulfanediide SMILES: [S--].[S--].[S--].[In+3].[In+3]
3
Indium(III) sulfide, 99.98% (metals basis)
CAS: 12030-24-9 Molecular Formula: In2S3 Molecular Weight (g/mol): 325.82 MDL Number: MFCD00011061 InChI Key: SIXIBASSFIFHDK-UHFFFAOYSA-N IUPAC Name: diindium(3+) trisulfanediide SMILES: [S--].[S--].[S--].[In+3].[In+3]