accessibility menu, dialog, popup

UserName

CAS RN 1202757-89-8

H3COONHNNNHNHOCH2F

CAS RN 1202757-89-8

IUPAC Name: N-{3-[(5-fluoro-2-{[4-(2-methoxyethoxy)phenyl]amino}pyrimidin-4-yl)amino]phenyl}prop-2-enamide
Synonyms: N-{3-[(5-fluoro-2-{[4-(2-methoxyethoxy)phenyl]amino}pyrimidin-4-yl)amino]phenyl}prop-2-enamide
Molecular Weight (g/mol): 423.45
Molecular Formula: C22H22FN5O3
InChi Key: KXBDTLQSDKGAEB-UHFFFAOYSA-N
SMILES: COCCOC1=CC=C(NC2=NC=C(F)C(NC3=CC(NC(=O)C=C)=CC=C3)=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results