accessibility menu, dialog, popup

UserName

BFe

CAS RN 12006-84-7

IUPAC Name: ferrioborane
Synonyms: iron boride
Molecular Weight (g/mol): 66.66
Molecular Formula: BFe
InChi Key: UJYBTTFGLAOKMN-UHFFFAOYSA-N
SMILES: [B+]=[Fe-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results