accessibility menu, dialog, popup

UserName

CAS RN 120011-70-3

IUPAC Name: hydrogen 2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one chloride
Synonyms: donepezil hydrochloride donepezil hcl
Molecular Weight (g/mol): 415.96
Molecular Formula: C24H30ClNO3
InChi Key: XWAIAVWHZJNZQQ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC1=CC2=C(C=C1OC)C(=O)C(CC1CCN(CC3=CC=CC=C3)CC1)C2
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)