CAS RN 12001-26-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mica, Water Ground, 325 Mesh, Spectrum™ Chemical
CAS: 12001-26-2 Molecular Weight (g/mol): 797 g/mol