accessibility menu, dialog, popup

UserName

HN

CAS RN 120-72-9

IUPAC Name: 1H-indole
Synonyms: indole 2,3-benzopyrrole 14c-labeled indole ketole caswell no. 498B indole, 14C-labeled 14C-labeled indole indole (substance) indole, 14c-labeled epa pesticide chemical code 025000
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: SIKJAQJRHWYJAI-UHFFFAOYSA-N
SMILES: N1C=CC2=CC=CC=C12
Boiling Point 
  • (3)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (3)
  • (3)
  • (4)
  • (4)
  • (6)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (7)