CAS RN 1199-01-5
IUPAC Name:
2-phenyl-4,5-dihydro-1,3-oxazol-5-one
Synonyms:
phenyloxazolone 2-phenyloxazolone
Molecular Weight (g/mol):
161.16
Molecular Formula:
C9H7NO2
InChi Key:
QKCKCXFWENOGER-UHFFFAOYSA-N
SMILES:
O=C1CN=C(O1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Phenyl-5-oxazolone, 97%
2-Phenyl-5-oxazolone, 97%
CAS: 1199-01-5 Molecular Formula: C9H7NO2 Molecular Weight (g/mol): 161.16 MDL Number: MFCD00014517 InChI Key: QKCKCXFWENOGER-UHFFFAOYSA-N Synonym: 2-phenyloxazol-5 4h-one,phenyloxazolone,2-phenyl-5-oxazolone,2-phenyloxazolone,2-phenyloxazolin-5-one,5 4h-oxazolone, 2-phenyl,2-phenyl-5 4h-oxazolone,2-phenyl-1,3-oxazol-5 4h-one,2-phenyl-4h-oxazol-5-one,2-oxazolin-5-one, 2-phenyl PubChem CID: 65073 ChEBI: CHEBI:60296 IUPAC Name: 2-phenyl-4H-1,3-oxazol-5-one SMILES: C1C(=O)OC(=N1)C2=CC=CC=C2