CAS RN 119670-30-3
CAS RN 119670-30-3
IUPAC Name:
benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate
Synonyms:
benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate
Molecular Weight (g/mol):
445.52
Molecular Formula:
C22H31N5O5
InChi Key:
ZLDBPOCJCXPVND-OALUTQOASA-N
SMILES:
CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)NC(=O)[C@@H](NCC(=O)C=[N+]=[N-])C(C)C
1
–
1
of
1
results
1
Thermo Scientific Chemicals Z-Leu-Val-Gly Diazomethyl Ketone
CAS: 119670-30-3 Molecular Formula: C22H31N5O5,C22H31N5O5 Molecular Weight (g/mol): 445.52 InChI Key: ZLDBPOCJCXPVND-OALUTQOASA-N Synonym: C-3455 IUPAC Name: benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate SMILES: CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)NC(=O)[C@@H](NCC(=O)C=[N+]=[N-])C(C)C