accessibility menu, dialog, popup

UserName

H3COCH3HO

CAS RN 1195-09-1

IUPAC Name: 2-methoxy-5-methylphenol
Synonyms: 2-methoxy-5-methylphenol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: IFNDEOYXGHGERA-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results