accessibility menu, dialog, popup

UserName

CAS RN 119313-12-1

IUPAC Name: 2-benzyl-2-(dimethylamino)-1-[4-(morpholin-4-yl)phenyl]butan-1-one
Synonyms: benzyl dimethylamino morpholinophenyl butanone
Molecular Weight (g/mol): 366.51
Molecular Formula: C23H30N2O2
InChi Key: UHFFVFAKEGKNAQ-UHFFFAOYNA-N
SMILES: CCC(CC1=CC=CC=C1)(N(C)C)C(=O)C1=CC=C(C=C1)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results