CAS RN 1190380-49-4
CAS RN 1190380-49-4
IUPAC Name:
1-(oxan-4-yl)-1H-pyrazol-4-amine
Synonyms:
1-(oxan-4-yl)pyrazol-4-amine
Molecular Weight (g/mol):
167.21
Molecular Formula:
C8H13N3O
InChi Key:
GZZNBQLBGSVOOZ-UHFFFAOYSA-N
SMILES:
NC1=CN(N=C1)C1CCOCC1
1
–
1
of
1
results
1
1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine, 97%, Thermo Scientific™
CAS: 1190380-49-4 Molecular Formula: C8H13N3O Molecular Weight (g/mol): 167.21 MDL Number: MFCD11505007 InChI Key: GZZNBQLBGSVOOZ-UHFFFAOYSA-N Synonym: 1-tetrahydro-2h-pyran-4-yl-1h-pyrazol-4-amine,1-oxan-4-yl pyrazol-4-amine,1-oxan-4-yl-1h-pyrazol-4-amine,1h-pyrazol-4-amine, 1-tetrahydro-2h-pyran-4-yl,1-tetrahydropyran-4-yl pyrazol-4-amine,1-tetrahydro-pyran-4-yl-1h-pyrazol-4-ylamine,1-2h-3,4,5,6-tetrahydropyran-4-yl pyrazole-4-ylamine PubChem CID: 51063703 IUPAC Name: 1-(oxan-4-yl)pyrazol-4-amine SMILES: NC1=CN(N=C1)C1CCOCC1